Oxamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11570-0E A11570-22 A11570-36
CAAAA11570-0EEA 970.12 CAD
CAAAA11570-0E CAAAA11570-22 CAAAA11570-36
Oxamide 98+%
Oxamide

MDL: MFCD00008007 Beilstein Registry No.: 1743262


Purity: 98+% CAS No.: 471-46-5 Molecular Formula: C2H4N2O2 Formula Weight: 88.07 Odor: Odorless Melting Point: ca 350° dec. Beilstein Reference: 1743262 Merck Reference: 14,6918 MDL No.: MFCD00008007

Formula: C₂H₄N₂O₂
Melting Pt: ca 350 °C (dec.)
Density: 1.667
Storage Temperature: Ambient
MDL Number: MFCD00008007
CAS Number: 471-46-5
EINECS: 207-442-5
Merck Index: 14,6918

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR