4-Amino-2,6-dichlorophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dichloro-4-hydroxyaniline

A11568-30 A11568-09 A11568-18
CAAAAA11568-30EA 551.42 CAD
CAAAAA11568-30 CAAAA11568-09 CAAAA11568-18
4-Amino-2,6-dichlorophenol 97%
4-Amino-2,6-dichlorophenol

MDL: MFCD00007875 Beilstein Registry No.: 2361665


Purity: 97% CAS No.: 5930-28-9 Molecular Formula: C6H5Cl2NO Formula Weight: 178.02 Melting Point: 164-169° Beilstein Reference: 2361665

Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 164…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007875
CAS Number: 5930-28-9
EINECS: 227-671-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR