α-Chloro-2-fluorotoulene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11566-14
A11566-22
CAAAA11566-14EA
80.9
CAD
CAAAA11566-14
CAAAA11566-22
α-Chloro-2-fluorotoulene 98+%
α-Chloro-2-fluorotoulene
MDL: MFCD00000892
Beilstein Registry No.: 471699
Purity: 98+%
CAS No.: 345-35-7
Molecular Formula: C7H6ClF
Formula Weight: 144.58
Flash Point: 57°(134°F)
Refractive Index: 1.5154
Odor: Pungent
Boiling Point: 175°
Beilstein Reference: 471699
UN No.: 2920
MDL No.: MFCD00000892
Formula:
C₇H₆ClF Boiling Pt: 175 °C Density: 1.216 Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000892 CAS Number: 345-35-7 EINECS: 206-460-0 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...