p-Toluamide 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methylbenzamide

A11559-14 A11559-22 A11559-36
CAAAA11559-14EA 54.19 CAD
CAAAA11559-14 CAAAA11559-22 CAAAA11559-36
p-Toluamide 98+%
p-Toluamide

MDL: MFCD00008002 Beilstein Registry No.: 774353


Purity: 98+% CAS No.: 619-55-6 Molecular Formula: C8H9NO Formula Weight: 135.17 Melting Point: 158-160° Beilstein Reference: 774353

Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00008002
CAS Number: 619-55-6
EINECS: 210-600-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR