3,4-Dihydroxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Protocatechuic aldehyde, Protocatechualdehyde

A11558-06 A11558-14 A11558-22
CAAAA11558-06EA 84.21 CAD
CAAAA11558-06 CAAAA11558-14 CAAAA11558-22
3,4-Dihydroxybenzaldehyde 98%
3,4-Dihydroxybenzaldehyde

MDL: MFCD00003370 Beilstein Registry No.: 774381


Purity: 98% CAS No.: 139-85-5 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 152-157° Beilstein Reference: 774381 Merck Reference: 14,7893 MDL No.: MFCD00003370

Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 295
Melting Pt: 152…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00003370
CAS Number: 139-85-5
EINECS: 205-377-7
Merck Index: 14,7893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR