N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: ACES, N-(Carbamoylmethyl)-2-aminoethanesulfonic Acid, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulfonic acid

A11553-14 A11553-22 A11553-36
CAAAA11553-14EA 83.27 CAD
CAAAA11553-14 CAAAA11553-22 CAAAA11553-36
N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) 99%
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)

Good's buffers


Purity: 99% CAS No.: 7365-82-4 Molecular Formula: H2NCOCH2NHCH2CH2SO3H Formula Weight: 182.20 Melting Point: ca 275° dec. Beilstein Reference: 2253770 Merck Reference: 14,36 MDL No.: MFCD00008030

Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182.2 g/mol
Melting Pt: ∼ 275 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008030
CAS Number: 7365-82-4
EINECS: 230-908-4
Merck Index: 14,36
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR