2',4',6'-Trimethylacetophenone 97+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(2,4,6-Trimethylphenyl)ethan-1-one

A11549-14 A11549-22
CAAAA11549-14EA 155.95 CAD
CAAAA11549-14 CAAAA11549-22
2',4',6'-Trimethylacetophenone 97+%
2',4',6'-Trimethylacetophenone

MDL: MFCD00008735 Beilstein Registry No.: 2044595


Purity: 97+% CAS No.: 1667-01-2 Molecular Formula: C11H14O Formula Weight: 162.23 Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 235-236° Beilstein Reference: 2044595 MDL No.: MFCD00008735

Formula: C₁₁H₁₄O
Boiling Pt: 235…236 °C
Density: 0.975
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008735
CAS Number: 1667-01-2
EINECS: 216-783-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR