1,3-Diacetoxybenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11547-18
A11547-30
CAAAA11547-0BEA
535.32
CAD
CAAAA11547-0B
CAAAA11547-18
CAAAA11547-30
1,3-Diacetoxybenzene 98%
1,3-Diacetoxybenzene
MDL: MFCD00008701
Beilstein Registry No.: 1875007
Purity: 98%
CAS No.: 108-58-7
Molecular Formula: C10H10O4
Formula Weight: 194.19
Flash Point:
Formula:
C₁₀H₁₀O₄ Boiling Pt: 278 °C Melting Pt: 142…143 °C Density: 1,179 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008701 CAS Number: 108-58-7 EINECS: 203-596-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...