(±)-2-Methylbutyric acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11546-AC
A11546-0F
A11546-AP
CAAAAA11546-ACEA
40.45
CAD
CAAAAA11546-AC
CAAAA11546-0F
CAAAA11546-AP
(±)-2-Methylbutyric acid 98%
(±)-2-Methylbutyric acid
MDL: MFCD00002669
Beilstein Registry No.: 1720486
Purity: 98%
CAS No.: 116-53-0
Molecular Formula: C5H10O2
Formula Weight: 102.14
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 0.937
Refractive Index: 1.4055
Boiling Point: 175-176°
Beilstein Reference: 1720486
UN No.: 3265
MDL No.: MFCD00002669
Formula:
C₅H₁₀O₂ Boiling Pt: 175…176 °C Melting Pt: –70 °C Density: 0.937 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002669 CAS Number: 116-53-0 EINECS: 204-145-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...