Quinoline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00006736
Beilstein Registry No.: 107477
Notes: Dissolves sulfur, As2O3, phosphorus
More soluble in hot than cold water. Miscible with alcohol, ether
Fieser: 1,975 2,356 9,403
Purity: 97%
CAS No.: 91-22-5
Molecular Formula: C9H7N
Formula Weight: 129.16
Flash Point: 101°(214°F)
Refractive Index: 1.6260
Melting Point: -15°
Boiling Point: 237°
Solubility: More soluble in hot than cold water. Miscible with alcohol, ether
Beilstein Reference: 107477
Merck Reference: 14,8068
UN No.: 2656
MDL No.: MFCD00006736
Formula:
C₉H₇N Boiling Pt: 237 °C Melting Pt: –15 °C Density: 1.090 Flash Pt: 101 °C (214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006736 CAS Number: 91-22-5 EINECS: 202-051-6 UN: 2656 ADR: 6.1,III Merck Index: 14,8068 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...