Pyridine-3,5-dicarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dinicotinic acid, 3,5-Pyridinedicarboxylic acid

A11540-09 A11540-18
CAAAA11540-09EA 159.58 CAD
CAAAA11540-09 CAAAA11540-18
Pyridine-3,5-dicarboxylic acid 98%
Pyridine-3,5-dicarboxylic acid

MDL: MFCD00006393 Beilstein Registry No.: 131640


Purity: 98% CAS No.: 499-81-0 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: ca 325° dec. Beilstein Reference: 131640

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
Melting Pt: ∼ 325 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006393
CAS Number: 499-81-0
EINECS: 207-893-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR