Tetraphenylphosphonium iodide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11539-06 A11539-14 A11539-22
CAAAA11539-06EA 59.72 CAD
CAAAA11539-06 CAAAA11539-14 CAAAA11539-22
Tetraphenylphosphonium iodide 98+%
Tetraphenylphosphonium iodide

MDL: MFCD00011917 Beilstein Registry No.: 3922216


Purity: 98+% CAS No.: 2065-67-0 Molecular Formula: (C6H5)4PI Formula Weight: 466.31 Melting Point: 345-350° Beilstein Reference: 3922216

Formula: C₂₄H₂₀IP
Melting Pt: 345…350 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011917
CAS Number: 2065-67-0
EINECS: 218-179-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR