Molecular sieve A4 (0.4 nm, 4 Å), powder

Supplier: Thermo Scientific Chemicals

Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

A11535-0B A11535-0I A11535-30
CAAAA11535-0BEA 176.96 CAD
CAAAA11535-0B CAAAA11535-0I CAAAA11535-30
Molecular sieve A4 (0.4 nm, 4 Å), powder
Molecular sieve A4 (0.4 nm, 4 Å)

MDL: MFCD00131613


CAS No.: 70955-01-0 MDL No.: MFCD00131613

Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR