(4-Aminophenyl)acetic acid ethyl ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 4-aminophenylacetate, Ethyl (4-aminophenyl)acetate

A11529-06 A11529-14
CAAAA11529-06EA 142.36 CAD
CAAAA11529-06 CAAAA11529-14
(4-Aminophenyl)acetic acid ethyl ester 98%
(4-Aminophenyl)acetic acid ethyl ester

MDL: MFCD00017569


Purity: 98% CAS No.: 5438-70-0 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 46-48° MDL No.: MFCD00017569

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00017569
CAS Number: 5438-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR