Ethyl trifluoroacetate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoroacetic ethyl ester

A11520-0B A11520-18 A11520-30
CAAAA11520-0BEA 1401.46 CAD
CAAAA11520-0B CAAAA11520-18 CAAAA11520-30
Ethyl trifluoroacetate 99%
Ethyl trifluoroacetate

MDL: MFCD00000419 Beilstein Registry No.: 1761411 Fieser: 19,144


Purity: 99% CAS No.: 383-63-1 Molecular Formula: C4H5F3O2 Formula Weight: 142.08 Flash Point: -1°(30°F) Refractive Index: 1.3070 Odor: Sweet Melting Point: -78° Boiling Point: 60-62° Beilstein Reference: 1761411 UN No.: 2924 MDL No.: MFCD00000419

Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
Boiling Pt: 60…62 °C
Melting Pt: < –78 °C
Density: 1.192
Flash Pt: –1 °C (30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000419
CAS Number: 383-63-1
EINECS: 206-851-6
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR