4-Pentylphenol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Amylphenol
A11514-06
A11514-14
CAAAA11514-06EA
85
CAD
CAAAA11514-06
CAAAA11514-14
CAAAA11514-22
4-Pentylphenol 98%
4-Pentylphenol
MDL: MFCD00020211
Beilstein Registry No.: 1364424
Purity: 98%
CAS No.: 14938-35-3
Molecular Formula: C11H16O
Formula Weight: 164.25
Flash Point: 133°(271°F)
Refractive Index: 1.5145
Melting Point: 24°
Boiling Point: 342°
Beilstein Reference: 1364424
UN No.: 2430
MDL No.: MFCD00020211
Formula:
C₁₁H₁₆O MW: 164.25 g/mol Boiling Pt: 342 °C Melting Pt: 24 °C Density: 0.960 Flash Pt: 133 °C (271 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020211 CAS Number: 14938-35-3 EINECS: 239-015-4 UN: 2430 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...