trans-2-Hexenal 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11513-14
A11513-22
CAAAA11513-14EA
60.99
CAD
CAAAA11513-14
CAAAA11513-22
CAAAA11513-36
trans-2-Hexenal 96%
trans-2-Hexenal
MDL: MFCD00007008
Beilstein Registry No.: 1699684
Purity: 96%
CAS No.: 6728-26-3
Molecular Formula: C6H10O
Formula Weight: 98.15
Storage Temperature: 2°C to 8°C
Flash Point: 38°(100°F)
Refractive Index: 1.4470
Odor: Sweet
Boiling Point: 37-38°/8mm
Beilstein Reference: 1699684
UN No.: 1988
MDL No.: MFCD00007008
Formula:
C₆H₁₀O Boiling Pt: 37…38 °C/8 mm Density: 0.848 Flash Pt: 38 °C (100 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007008 CAS Number: 6728-26-3 EINECS: 229-778-1 UN: 1988 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...