4-Chlorobenzyl alcohol 99%
Supplier: Thermo Scientific Chemicals
A11504-14
A11504-22
CAAAA11504-14EA
104.91
CAD
CAAAA11504-14
CAAAA11504-22
CAAAA11504-36
4-Chlorobenzyl alcohol 99%
4-Chlorobenzyl alcohol
MDL: MFCD00004652
Beilstein Registry No.: 636502
Fieser: 7,59
Purity: 99%
CAS No.: 873-76-7
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Melting Point: 68-72°
Boiling Point: 234°
MDL No.: MFCD00004652
Formula:
C₇H₇ClO Boiling Pt: 234 °C Melting Pt: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004652 CAS Number: 873-76-7 EINECS: 212-852-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...