4-Bromocinnamic acid predominantly trans 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11503-22 A11503-06 A11503-14
CAAAAA11503-22EA 508.76 CAD
CAAAAA11503-22 CAAAA11503-06 CAAAA11503-14
4-Bromocinnamic acid predominantly trans 98+%
4-Bromocinnamic acid

MDL: MFCD00004394 Beilstein Registry No.: 3538823

Formula: C₉H₇BrO₂
Melting Pt: 260…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00004394
CAS Number: 1200-07-3
EINECS: 214-850-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR