2-Nitrobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11501-14 A11501-22 A11501-36
CAAAA11501-14EA 101.59 CAD
CAAAA11501-14 CAAAA11501-22 CAAAA11501-36
2-Nitrobenzaldehyde 98+%
2-Nitrobenzaldehyde

MDL: MFCD00007132 Beilstein Registry No.: 742624


Purity: 98+% CAS No.: 552-89-6 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Flash Point: 110°(230°F) Melting Point: 43-46° Boiling Point: 153°/23mm Beilstein Reference: 742624 Merck Reference: 14,6587 MDL No.: MFCD00007132

Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 153 °C/23 mm
Melting Pt: 43…46 °C
Density: 1.35 g/cm³ (25 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 14,6587

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR