p-Tolyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11499-22
A11499-06
A11499-14
CAAAAA11499-22EA
688.88
CAD
CAAAAA11499-22
CAAAA11499-06
CAAAA11499-14
p-Tolyl isothiocyanate 97%
p-Tolyl isothiocyanate
MDL: MFCD00004813
Beilstein Registry No.: 386032
Purity: 97%
CAS No.: 622-59-3
Molecular Formula: C8H7NS
Formula Weight: 149.22
Flash Point: 107°(224°F)
Refractive Index: 1.6345
Odor: Pungent
Melting Point: 23-26°
Boiling Point: 236-238°
Beilstein Reference: 386032
UN No.: 2923
MDL No.: MFCD00004813
Formula:
C₈H₇NS Boiling Pt: 236…238 °C Melting Pt: 23…26 °C Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004813 CAS Number: 622-59-3 EINECS: 210-745-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...