Ethyl cyanoacetate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11498-0E A11498-36
CAAAA11498-0EEA 312.84 CAD
CAAAA11498-0E CAAAA11498-36
Ethyl cyanoacetate 98+%
Ethyl cyanoacetate

MDL: MFCD00001940 Beilstein Registry No.: 605871


Purity: 98+% CAS No.: 105-56-6 Molecular Formula: C5H7NO2 Formula Weight: 113.12 Flash Point: 109°(228°F) Refractive Index: 1.4180 Odor: Weak Melting Point: -23° Boiling Point: 208-210° Beilstein Reference: 605871 Merck Reference: 14,3786 MDL No.: MFCD00001940

Formula: C₅H₇NO₂
MW: 113.12 g/mol
Boiling Pt: 209
Melting Pt: -22
Density: 1.063
Flash Pt: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001940
CAS Number: 105-56-6
EINECS: 203-309-0
Merck Index: 14,3786

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR