1-Chlorobutane 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Butyl Chloride

A11494-0F A11494-AP
CAAAAA11494-AEEA 214.64 CAD
CAAAAA11494-AE CAAAA11494-0F CAAAA11494-AP
1-Chlorobutane 99+%
1-Chlorobutane

MDL: MFCD00001009 Beilstein Registry No.: 1730909


Purity: 99+% CAS No.: 109-69-3 Molecular Formula: CH3(CH2)3Cl Formula Weight: 92.57 Flash Point: -12°(10°F) Physical Form: Liquid Density: 0.886 Refractive Index: 1.4024 Odor: Pungent Melting Point: -123° Boiling Point: 77-78° Beilstein Reference: 1730909 Merck Reference: 14,1560 UN No.: 1127 MDL No.: MFCD00001009

Formula: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Boiling Pt: 77…78 °C
Melting Pt: –123 °C
Density: 0.886
Flash Pt: –12 °C (10 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3 II
Merck Index: 14,1560
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR