3-Iodo-p-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11485-06
A11485-14
A11485-22
CAAAA11485-06EA
65.1
CAD
CAAAA11485-06
CAAAA11485-14
CAAAA11485-22
3-Iodo-p-toluidine 98%
3-Iodo-p-toluidine
MDL: MFCD00047843
Beilstein Registry No.: 2353567
Purity: 98%
CAS No.: 35944-64-0
Molecular Formula: C7H8IN
Formula Weight: 233.05
Melting Point: 34-37°
Beilstein Reference: 2353567
UN No.: 2811
MDL No.: MFCD00047843
Formula:
C₇H₈IN Melting Pt: 34…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047843 CAS Number: 35944-64-0 EINECS: 252-803-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...