Chloroacetic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A11482-0B A11482-30
CAAAA11482-0BEA 167.48 CAD
CAAAA11482-0B CAAAA11482-30
Chloroacetic acid 99%
Chloroacetic acid

MDL: MFCD00002683 Beilstein Registry No.: 605438 Soluble in water, alcohols, benzene, chloroform, ether


Purity: 99% CAS No.: 79-11-8 Molecular Formula: C2H3ClO2 Formula Weight: 94.50 Flash Point: 126°(258°F) Odor: Pungent pH: 1.0 Melting Point: 60-63° Boiling Point: 189° Solubility: Soluble in water, alcohols, benzene, chloroform, ether Beilstein Reference: 605438 Merck Reference: 14,2112 UN No.: 1751 MDL No.: MFCD00002683

Formula: ClCH₂COOH
Boiling Pt: 189 °C
Melting Pt: 60…63 °C
Density: 1.404
Flash Pt: 126 °C (258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1,II
Merck Index: 14,2112
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2516e0c26c6c26f4f6f3656bc5b4256e7cf08207&vid=9c9b7ad094469d9089eb2f0ce88275d1423c2b46&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR