4,4'-Dimethoxybenzophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(4-methoxyphenyl)methanone

A11466-14 A11466-22 A11466-36
CAAAA11466-14EA 132.88 CAD
CAAAA11466-14 CAAAA11466-22 CAAAA11466-36
4,4'-Dimethoxybenzophenone 98+%
4,4'-Dimethoxybenzophenone

MDL: MFCD00008404 Beilstein Registry No.: 1878026


Purity: 98+% CAS No.: 90-96-0 Molecular Formula: C15H14O3 Formula Weight: 242.27 Melting Point: 142-146° Beilstein Reference: 1878026

Formula: C₁₅H₁₄O₃
Boiling Pt: 180 - 240 °C (13 -rr)
Melting Pt: 142…146 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008404
CAS Number: 90-96-0
EINECS: 202-028-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR