1-Bromo-4-propoxybenzene 98%

Supplier: Thermo Scientific Chemicals

A11462-06 A11462-14
CAAAA11462-06EA 113.13 CAD
CAAAA11462-06 CAAAA11462-14
1-Bromo-4-propoxybenzene 98%
1-Bromo-4-propoxybenzene

MDL: MFCD00156160


Purity: 98% CAS No.: 39969-56-7 Molecular Formula: C9H11BrO Formula Weight: 215.10 Flash Point: 110°(230°F) Boiling Point: 238-240° MDL No.: MFCD00156160

Formula: C₉H₁₁BrO
Boiling Pt: 238…240 °C
Density: 1,356
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00156160
CAS Number: 39969-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR