DL-Methionine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid
A11457-0B
A11457-0I
A11457-30
CAAAA11457-0BEA
167.48
CAD
CAAAA11457-0B
CAAAA11457-0I
CAAAA11457-30
DL-Methionine 99%
DL-Methionine
MDL: MFCD00063096
Beilstein Registry No.: 636185
Purity: 99%
CAS No.: 59-51-8
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Melting Point: ca 272° dec.
Beilstein Reference: 636185
Merck Reference: 14,5975
MDL No.: MFCD00063096
Formula:
C₅H₁₁NO₂S MW: 149.21 g/mol Melting Pt: ∼ 272 °C (dec.) Density: 1.34 Storage Temperature: Ambient |
MDL Number:
MFCD00063096 CAS Number: 59-51-8 EINECS: 200-432-1 Merck Index: 14,5975 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...