α,2,4-Trichlorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4-Dichlorobenzyl chloride, 1-(Chloromethyl)-2,4-dichlorobenzene, 1,3-Dichloro-4-(chloromethyl)benzene, 2,4-Dichloro-1-(chloromethyl)benzene
A11435-36
A11435-0B
CAAAAA11435-36EA
276.5
CAD
CAAAAA11435-36
CAAAA11435-0B
α,2,4-Trichlorotoluene 98%
α,2,4-Trichlorotoluene
MDL: MFCD00000895
Beilstein Registry No.: 387220
Purity: 98%
CAS No.: 94-99-5
Molecular Formula: C7H5Cl3
Formula Weight: 195.48
Flash Point: 190°(374°F)
Refractive Index: 1.5760
Odor: Pungent
Melting Point: -3°
Boiling Point: 248°
Beilstein Reference: 387220
UN No.: 3265
MDL No.: MFCD00000895
Formula:
C₇H₅Cl₃ MW: 195.47 g/mol Boiling Pt: 248 °C Melting Pt: –3 °C Density: 1.408 Flash Pt: 190 °C (374 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000895 CAS Number: 94-99-5 EINECS: 202-381-0 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...