2,5-Dichlorothiophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A11434-03 A11434-06 A11434-14
CAAAAA11434-03EA 38.08 CAD
CAAAAA11434-03 CAAAAA11434-22 CAAAA11434-06 CAAAA11434-14
2,5-Dichlorothiophenol 98%
2,5-Dichlorothiophenol

MDL: MFCD00004831 Beilstein Registry No.: 2042866


Purity: 98% CAS No.: 5858-18-4 Molecular Formula: C6H4Cl2S Formula Weight: 179.07 Flash Point: 110°(230°F) Refractive Index: 1.6190 Odor: Unpleasant Melting Point: 22-25° Boiling Point: 112-116°/50mm Beilstein Reference: 2042866 MDL No.: MFCD00004831

Formula: C₆H₄Cl₂S
MW: 179.07
Boiling Pt: 202
Melting Pt: 28
Density: 1.415
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004831
CAS Number: 5858-18-4
EINECS: 227-486-9
UN: 3335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR