5-Amino-1-naphthalenesulfonic acid tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Aminonaphthalene-1-sulfonic acid, 5-Aminonaphthalene-1-sulphonic acid, 5-Amino-α-naphthalenesulfonic acid, 5-Amino-α-naphthalenesulphonic acid

A11433-18 A11433-14 A11433-22
CAAAAA11433-18EA 113.29 CAD
CAAAAA11433-18 CAAAA11433-14 CAAAA11433-22
5-Amino-1-naphthalenesulfonic acid tech. 90%, Technical Grade
5-Amino-1-naphthalenesulphonic acid

MDL: MFCD00014315 Beilstein Registry No.: 2214149


Purity: 90% CAS No.: 84-89-9 Molecular Formula: C10H9NO3S Formula Weight: 223.25 Odor: Odorless Melting Point: 300° Beilstein Reference: 2214149 Merck Reference: 14,6404 MDL No.: MFCD00014315

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00014315
CAS Number: 84-89-9
EINECS: 201-571-0
Merck Index: 14,6404

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR