2',5'-Dimethylacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2,5-Dimethylphenyl)ethanone, 1-(2,5-Dimethylphenyl)ethan-1-one
A11432-06
A11432-14
CAAAA11432-06EA
52.14
CAD
CAAAA11432-06
CAAAA11432-14
CAAAA11432-22
2',5'-Dimethylacetophenone 98+%
2',5'-Dimethylacetophenone
MDL: MFCD00014988
Beilstein Registry No.: 2042139
Purity: 98+%
CAS No.: 2142-73-6
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 95°(203°F)
Refractive Index: 1.5290
Odor: Characteristic
Boiling Point: 230-231°
Beilstein Reference: 2042139
MDL No.: MFCD00014988
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 230…231 °C Melting Pt: –18 °C Density: 1.0154 g/cm³ (0 °C) Flash Pt: 95 °C(203 °F) |
MDL Number:
MFCD00014988 CAS Number: 2142-73-6 EINECS: 218-399-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...