Ethyl sorbate 97%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Ethyl trans,trans-hexa-2,4-dienoate, (2E,4E)-Ethyl hexa-2,4-dienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, 2,4-Hexadienoic acid ethyl ester, Sorbic acid ethyl ester, (2E,4E)-2,4-Hexadienoic acid ethyl ester, trans,trans-2,4-Hexadienoic acid ethyl ester, (E,E)-2,4-Hexadienoic acid ethyl ester, Ethyl (E,E)-hexa-2,4-dienoate
MDL: MFCD00009296
Beilstein Registry No.: 1362941
Purity: 98%
CAS No.: 2396-84-1
Molecular Formula: CH3CH=CHCH=CHCO2CH2CH3
Formula Weight: 140.18
Flash Point: 69°(156°F)
Refractive Index: 1.4950
Boiling Point: 195-196°
Beilstein Reference: 1362941
MDL No.: MFCD00009296
Formula:
C₈H₁₂O₂ MW: 140.18 g/mol Boiling Pt: 195.4 Density: 0.948 Flash Pt: 69 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009296 CAS Number: 2396-84-1 EINECS: 219-258-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...