4-Bromophenyl methyl sulfone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-4-(methylsulphonyl)benzene, 1-Bromo-4-(methylsulfonyl)benzene

A11427-09 A11427-18
CAAAA11427-09EA 80.26 CAD
CAAAA11427-09 CAAAA11427-18 CAAAA11427-30
4-Bromophenyl methyl sulfone 98+%
4-Bromophenyl methyl sulfone

MDL: MFCD00025065 Beilstein Registry No.: 2045675


Purity: 98+% CAS No.: 3466-32-8 Molecular Formula: C7H7BrO2S Formula Weight: 235.10 Melting Point: 101-102° Beilstein Reference: 2045675

Formula: C₇H₇BrO₂S
MW: 235.1 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00025065
CAS Number: 3466-32-8
EINECS: 222-421-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR