1,2,4-Trifluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11424-06
A11424-14
A11424-22
CAAAA11424-06EA
27.02
CAD
CAAAA11424-06
CAAAA11424-14
CAAAA11424-22
1,2,4-Trifluorobenzene 98+%
1,2,4-Trifluorobenzene
MDL: MFCD00000305
Beilstein Registry No.: 2042865
Purity: 98+%
CAS No.: 367-23-7
Molecular Formula: C6H3F3
Formula Weight: 132.09
Flash Point: 4°(39°F)
Refractive Index: 1.4230
Boiling Point: 89-91°
Beilstein Reference: 2042865
UN No.: 2387
MDL No.: MFCD00000305
Formula:
C₆H₃F₃ Boiling Pt: 89…91 °C Melting Pt: –35 °C Density: 1.264 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000305 CAS Number: 367-23-7 EINECS: 206-684-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...