1,1,3,3-Tetramethoxypropane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Malonaldehyde bis(dimethyl acetal)

A11418-0B A11418-0I A11418-30
CAAAA11418-0BEA 233.84 CAD
CAAAA11418-0B CAAAA11418-0I CAAAA11418-30
1,1,3,3-Tetramethoxypropane 98%
1,1,3,3-Tetramethoxypropane

MDL: MFCD00008488 Beilstein Registry No.: 1700020


Purity: 98+% CAS No.: 102-52-3 Molecular Formula: C7H16O4 Formula Weight: 164.20 Flash Point: 54°(129°F) Refractive Index: 1.4070 Boiling Point: 182-183° Beilstein Reference: 1700020 UN No.: 3271 MDL No.: MFCD00008488

Formula: C₇H₁₆O₄
Boiling Pt: 182…183 °C
Density: 0.997
Flash Pt: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008488
CAS Number: 102-52-3
EINECS: 203-037-2
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR