Bromobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenylbromide, Phenyl bromide

A11407-0C A11407-0E A11407-36
CAAAA11407-0CEA 791.58 CAD
CAAAA11407-0C CAAAA11407-0E CAAAA11407-36
Bromobenzene 99%
Bromobenzene

MDL: MFCD00000055 Beilstein Registry No.: 1236661


Purity: 99% CAS No.: 108-86-1 Molecular Formula: C6H5Br Formula Weight: 157.02 Flash Point: 51°(123°F) Physical Form: Liquid Density: 1.492 Refractive Index: 1.5590 Odor: Aromatic Melting Point: -31° Boiling Point: 155-156° Beilstein Reference: 1236661 Merck Reference: 14,1406 UN No.: 2514 MDL No.: MFCD00000055

Formula: C₆H₅Br
MW: 157.01 g/mol
Boiling Pt: 155
Melting Pt: -31
Density: 1.492
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000055
CAS Number: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3 III
Merck Index: 14,1406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR