α,α,α-Trifluoroacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11403-06
A11403-14
A11403-22
CAAAAA11403-06EA
110.13
CAD
CAAAAA11403-06
CAAAA11403-14
CAAAA11403-22
α,α,α-Trifluoroacetophenone 98%
α,α,α-Trifluoroacetophenone
MDL: MFCD00000420
Beilstein Registry No.: 1866286
Fieser: 1,1226 3,308
Purity: 98%
CAS No.: 434-45-7
Molecular Formula: C8H5F3O
Formula Weight: 174.12
Flash Point: 42°(108°F)
Refractive Index: 1.4595
Melting Point: -40°
Boiling Point: 165-166°
Beilstein Reference: 1866286
UN No.: 1224
MDL No.: MFCD00000420
Formula:
C₈H₅F₃O Boiling Pt: 165…166 °C Melting Pt: –40 °C Density: 1.240 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000420 CAS Number: 434-45-7 EINECS: 207-103-1 UN: 1993 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...