1-Bromo-4-chlorobutane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11399-14
A11399-22
CAAAA11399-14EA
75.52
CAD
CAAAA11399-14
CAAAA11399-22
1-Bromo-4-chlorobutane 99%
1-Bromo-4-chlorobutane
MDL: MFCD00001010
Beilstein Registry No.: 1098275
Purity: 99%
CAS No.: 6940-78-9
Molecular Formula: Br(CH2)4Cl
Formula Weight: 171.47
Flash Point: 60°(140°F)
Refractive Index: 1.4875
Odor: Aromatic
Boiling Point: 175-176°
Beilstein Reference: 1098275
UN No.: 1993
MDL No.: MFCD00001010
Formula:
BrH₂C(CH₂)₂CH₂Cl Boiling Pt: 175…176 °C Density: 1.489 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001010 CAS Number: 6940-78-9 EINECS: 230-089-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...