Neocuproine hemihydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11398-06
A11398-14
A11398-22
CAAAA11398-06EA
114.55
CAD
CAAAA11398-06
CAAAA11398-14
CAAAA11398-22
Neocuproine hemihydrate 98+%
Neocuproine hemihydrate
Reagent for the colorimetric determination of copper
Purity: 98+%
CAS No.: 34302-69-7
Molecular Formula: C14H12N2.0.5H2O
Formula Weight: 217.27 (208.26anhy)
Melting Point: 160-164°
Beilstein Reference: 153179
Merck Reference: 14,6449
MDL No.: MFCD00149306
Formula:
C₂₈H₂₆N₄O Melting Pt: 160…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149306 CAS Number: 34302-69-7 EINECS: 207-601-9 Merck Index: 14,6449 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...