3-Fluorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A11396-30 A11396-09 A11396-18
CAAAAA11396-30EA 838.98 CAD
CAAAAA11396-30 CAAAA11396-09 CAAAA11396-18
3-Fluorobenzaldehyde 97%
3-Fluorobenzaldehyde

MDL: MFCD00003348 Beilstein Registry No.: 970178


Purity: 97% CAS No.: 456-48-4 Molecular Formula: C7H5FO Formula Weight: 124.11 Flash Point: 56°(132°F) Refractive Index: 1.5190 Boiling Point: 173° Beilstein Reference: 970178 UN No.: 1989 MDL No.: MFCD00003348

Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 173 °C
Density: 1.174
Flash Pt: 56 °C (132 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR