4-Tolualdehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde

A11384-06 A11384-0E A11384-22
CAAAAA11384-06EA 53.56 CAD
CAAAAA11384-06 CAAAA11384-0E CAAAA11384-22
4-Tolualdehyde 98%
4-Tolualdehyde

MDL: MFCD00006954 Beilstein Registry No.: 385772


Purity: 98% CAS No.: 104-87-0 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 80°(176°F) Refractive Index: 1.5460 Melting Point: -6° Boiling Point: 204° Beilstein Reference: 385772 MDL No.: MFCD00006954

Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 204 °C
Melting Pt: –6 °C
Density: 1.018
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR