4-Chloro-3-nitrotoluene 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11383-22
A11383-36
CAAAA11383-0EEA
2978.96
CAD
CAAAA11383-0E
CAAAA11383-22
CAAAA11383-36
4-Chloro-3-nitrotoluene 97+%
4-Chloro-3-nitrotoluene
MDL: MFCD00007085
Beilstein Registry No.: 511055
Purity: 97+%
CAS No.: 89-60-1
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Flash Point: 129°(264°F)
Refractive Index: 1.5580
Melting Point: ca 7°
Boiling Point: 260-262°
Beilstein Reference: 511055
UN No.: 2433
MDL No.: MFCD00007085
Formula:
C₇H₆ClNO₂ Boiling Pt: 260…262 °C Melting Pt: ∼ 7 °C Density: 1.295 Flash Pt: 129 °C (264 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007085 CAS Number: 89-60-1 EINECS: 201-922-8 UN: 2433 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...