TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl
MDL: MFCD00012590
Beilstein Registry No.: 3675235
Purity: 99+%
CAS No.: 1185-53-1
Molecular Formula: C4H11NO3.HCl
Formula Weight: 157.60
pH: 3.5-5.0
Melting Point: ca 152° dec.
Beilstein Reference: 3675235
MDL No.: MFCD00012590
Formula:
NH₂C(CH₂OH)₃·HCl MW: 157.6 g/mol Melting Pt: ∼ 152 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012590 CAS Number: 1185-53-1 EINECS: 214-684-5 Merck Index: 12,09902 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...