4-Chloroanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

A11377-18 A11377-30
CAAAA11377-0BEA 2249.18 CAD
CAAAA11377-0B CAAAA11377-18 CAAAA11377-30
4-Chloroanisole 99%
4-Chloroanisole

MDL: MFCD00000616 Beilstein Registry No.: 1858901


Purity: 99% CAS No.: 623-12-1 Molecular Formula: C7H7ClO Formula Weight: 142.59 Flash Point: 78°(172°F) Refractive Index: 1.5350 Melting Point: -18° Boiling Point: 198-200° Beilstein Reference: 1858901 MDL No.: MFCD00000616

Formula: C₇H₇ClO
Boiling Pt: 198…200 °C
Melting Pt: –18 °C
Density: 1.170
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000616
CAS Number: 623-12-1
EINECS: 210-772-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR