2-Chloro-3-hydroxypyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-3-pyridinol
A11376-14
A11376-22
CAAAA11376-06EA
125.97
CAD
CAAAA11376-06
CAAAA11376-14
CAAAA11376-22
2-Chloro-3-hydroxypyridine 98%
2-Chloro-3-hydroxypyridine
MDL: MFCD00006235
Beilstein Registry No.: 2216
Purity: 98%
CAS No.: 6636-78-8
Molecular Formula: C5H4ClNO
Formula Weight: 129.55
Melting Point: 168-172°
Beilstein Reference: 2216
Formula:
C₅H₄ClNO MW: 129.55 g/mol Boiling Pt: 212 Melting Pt: 168…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006235 CAS Number: 6636-78-8 EINECS: 229-635-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...