Tetrahexylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

A11372-09 A11372-18 A11372-30
CAAAA11372-09EA 92.27 CAD
CAAAA11372-09 CAAAA11372-18 CAAAA11372-30
Tetrahexylammonium bromide 98%
Tetrahexylammonium bromide

MDL: MFCD00011858 Beilstein Registry No.: 3637302


Purity: 98% CAS No.: 4328-13-6 Molecular Formula: [CH3(CH2)5]4NBr Formula Weight: 434.60 Melting Point: ca 95-100° Beilstein Reference: 3637302

Formula: C₂₄H₅₂BrN
MW: 434.59 g/mol
Melting Pt: ∼ 95…100 °C
Density: 1.0036 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011858
CAS Number: 4328-13-6
EINECS: 224-363-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR