2,6-Dichloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

A11364-22 A11364-36
CAAAA11364-22EA 85.32 CAD
CAAAA11364-22 CAAAA11364-36
2,6-Dichloropyridine 98%
2,6-Dichloropyridine

MDL: MFCD00006244 Beilstein Registry No.: 108664


Purity: 98% CAS No.: 2402-78-0 Molecular Formula: C5H3Cl2N Formula Weight: 147.99 Odor: Unpleasant Melting Point: 84-88° Boiling Point: 211° Beilstein Reference: 108664 UN No.: 2811 MDL No.: MFCD00006244

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 211 °C
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006244
CAS Number: 2402-78-0
EINECS: 219-282-3
UN: 2811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR