α,α,α,α',α',α'-Hexafluoro-m-xylene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene
A11360-0B
A11360-18
A11360-30
CAAAA11360-0BEA
848.46
CAD
CAAAA11360-0B
CAAAA11360-18
CAAAA11360-30
α,α,α,α',α',α'-Hexafluoro-m-xylene 98+%
α,α,α,α',α',α'-Hexafluoro-m-xylene
MDL: MFCD00000392
Beilstein Registry No.: 2052589
Purity: 98+%
CAS No.: 402-31-3
Molecular Formula: C8H4F6
Formula Weight: 214.11
Flash Point: 26°(78°F)
Refractive Index: 1.3790
Odor: Aromatic
Melting Point: -35°
Boiling Point: 115-117°
Beilstein Reference: 2052589
UN No.: 1993
MDL No.: MFCD00000392
Formula:
C₈H₄F₆ Boiling Pt: 115…117 °C Melting Pt: –35 °C Density: 1.378 Flash Pt: 26 °C (78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000392 CAS Number: 402-31-3 EINECS: 206-939-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...