2-Amino-5-nitropyridine ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11352-14
A11352-22
CAAAA11352-14EA
139.83
CAD
CAAAA11352-14
CAAAA11352-22
CAAAA11352-36
2-Amino-5-nitropyridine ≥97%
2-Amino-5-nitropyridine
MDL: MFCD00006325
Beilstein Registry No.: 120353
Purity: 97%
CAS No.: 4214-76-0
Molecular Formula: C5H5N3O2
Formula Weight: 139.11
Flash Point: 224°(435°F)
Melting Point: 185-190°
Beilstein Reference: 120353
MDL No.: MFCD00006325
Formula:
C₅H₅N₃O₂ Melting Pt: 185…190 °C Density: 1.623 g/cm³ (20 °C) Flash Pt: 224 °C(435 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006325 CAS Number: 4214-76-0 EINECS: 224-145-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...